EMAIL THIS PAGE TO A FRIEND
Molecular medicine (Cambridge, Mass.)
Is there a role for high mobility group box 1 and the receptor for advanced glycation end products in the genesis of long-term cognitive impairment in sepsis survivors?
PMID 23114886