EMAIL THIS PAGE TO A FRIEND
Free radical research
Hydrogen peroxide induces overexpression of angiotensin-converting enzyme in human umbilical vein endothelial cells.
PMID 23153326
Abstract
Oxidative stress has been linked to endothelial dysfunction in atherosclerosis and hypertension. The present study was designed to investigate the effect of hydrogen peroxide (H2O2) on angiotensin-converting enzyme (ACE), a key regulator of the renin-angiotensin system, and the mechanisms underlying ACE regulation in human umbilical vein endothelial cells (HUVECs). We used Tetrazolium bromide (MTT) assay for cell viability, terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling (TUNEL) assay for cell apoptosis, enzyme-linked immunosorbent assay (ELISA) for cAMP measurement, real-time PCR for mRNA detection, and Western blot for protein analysis in the study. Our results demonstrated that H2O2 (50-1000 μM) decreased HUVECs viability by inducing apoptosis. Notably, H2O2 upregulated ACE expression in a concentration-dependent manner. H2O2 100 μM significantly enhanced cyclic adenosine monophosphate (cAMP) expression by 1.48-fold (P<0.05). Additionally, forskolin 10 μM, a cAMP agonist, was also found to enhance ACE expression by 1.78-fold (P<0.05); in contrast, H-89 10 μM, a protein kinase A (PKA) inhibitor, abolished H2O2-induced ACE expression and prevented the enhancing effect of forskolin-induced ACE expression. Similar effects on ACE mRNA were also observed. cAMP-response element-specific decoy oligodeoxynucleotides (CRE-dODN) containing binding sites for cAMP-response element-binding protein (CREB) inhibited ACE expression at both the mRNA and protein levels. Negative control CRE-dODN had no effect on ACE expression. We conclude that H2O2 upregulates the expression of ACE through the activation of cAMP/PKA/CREB signal pathway in HUVECs, indicating a role of oxidative stress in the pathophysiology of hypertension.