EMAIL THIS PAGE TO A FRIEND
FEBS letters
Calmodulin activates neuronal nitric oxide synthase by enabling transitions between conformational states.
PMID 23159936
Abstract
We recently showed that inducible nitric oxide synthase conformational intermediates can be resolved via FMN fluorescence lifetimes. Here we show that neuronal NOS activation by calmodulin removes constraints favoring a closed 'input state', increasing occupation of other states and facilitating conformational transitions. The 90 ps FMN input state lifetime distinguishes it from ∼4 ns 'open' states in which FMN does not interact strongly with other groups, or 0.9 ns output states in which FMN interacts with ferriheme. Enablement of the conformational cycle is an important paradigm for control in nNOS and related enzymes, and may extend to other control modalities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4915 Nitric Oxide Synthase Neuronal Human, recombinant, expressed in E. coli