EMAIL THIS PAGE TO A FRIEND
Reproduction, fertility, and development
Artificial fertilisation in a terrestrial toadlet (Pseudophryne guentheri): effect of medium osmolality, sperm concentration and gamete storage.
PMID 23174151
Abstract
Anurans exhibit a greater reproductive diversity than any other vertebrate order. However, studies investigating the effects of the external fertilisation environment on fertilisation success are limited to aquatic-breeding species. This study investigated the effects of fertilisation medium osmolality, sperm concentration and short-term oocyte storage on fertilisation success in a terrestrial-breeding anuran, Pseudophryne guentheri. Split-clutch experimental designs were used to determine optimal fertilisation conditions. To determine the effect of short-term sperm storage, sperm viability was assessed using fluorescence microscopy and percentage sperm motility and velocity quantified with a computer-assisted sperm analysis system. Fertilisation success was highest in media ranging in osmolality from 25 mOsm kg⁻¹ to 100 mOsm kg⁻¹, representing a broader range and higher optimal osmolality than previously reported for aquatic breeders. High rates of fertilisation (>75%) were achieved in relatively low sperm concentrations (2.5×10⁴ mL⁻¹). Oocytes stored in isotonic solutions (200 mOsm kg⁻¹) retained fertilisation capacity (32%) after 8h of storage, while sperm suspensions maintained motility (≥26%) for 13 days. Additional studies on terrestrial-breeding anurans will be required to ascertain whether the optimal fertilisation conditions reported reflect adaptations to achieve fertilisation in a terrestrial environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0399 Leuprolide acetate salt, ≥98% (HPLC)
C59H84N16O12 · xC2H4O2
L0376000 Leuprorelin, European Pharmacopoeia (EP) Reference Standard
C59H84N16O12