EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Molecular signaling involved in oxysterol-induced β₁-integrin over-expression in human macrophages.
PMID 23203064
Abstract
The hypercholesterolemia-atherosclerosis association is now established; hypercholesterolemia may induce vascular-cell activation, subsequently increasing expression of adhesion molecules, cytokines, chemokines, growth factors, and other key inflammatory molecules. Among inflammatory molecules expressed by vascular cells, integrins play a critical role in regulating macrophage activation and migration to the site of inflammation, by mediating cell-cell and cell-extracellular matrix interactions. The main lipid oxidation products present in oxidized LDL that may be responsible for inflammatory processes in atherogenesis, are cholesterol oxidation products, known as oxysterols. This study demonstrates the effect of an oxysterol mixture, compatible with that detectable in human hypercholesterolemic plasma, on the expression and synthesis of β(1)-integrin in cells of the macrophage lineage. The molecular signaling whereby oxysterols induce β(1)-integrin up-regulation is also comprehensively investigated. Over-expression of β(1)-integrin depends on activation of classic and novel members of protein kinase C and extracellular signal-regulated kinases 1 and 2, as well as of the up-stream G-protein (Gq and G13), c-Src, and phospholipase C. In addition, the localization of β(1)-integrin in advanced human carotid plaques is highlighted, marking its importance in atherosclerotic plaque progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU011371 MISSION® esiRNA, esiRNA targeting mouse Map2k2 (esiRNA1)