PloS one
Phytochemicals attenuating aberrant activation of β-catenin in cancer cells.
PMID 23226522
Abstract
Phytochemicals are a rich source of chemoprevention agents but their effects on modulating the Wnt/β-catenin signaling pathway have remained largely uninvestigated. Aberrantly activated Wnt signaling can result in the abnormal stabilization of β-catenin, a key causative step in a broad spectrum of cancers. Here we report the modulation of lithium chloride-activated canonical Wnt/β-catenin signaling by phytochemicals that have antioxidant, anti-inflammatory or chemopreventive properties. The compounds were first screened with a cervical cancer-derived stable Wnt signaling reporter HeLa cell line. Positive hits were subsequently evaluated for β-catenin degradation, suppression of β-catenin nuclear localization and down-regulation of downstream oncogenic targets of Wnt/β-catenin pathway. Our study shows a novel degradation path of β-catenin protein in HeLa cells by Avenanthramide 2p (a polyphenol) and Triptolide (a diterpene triepoxide), respectively from oats and a Chinese medicinal plant. The findings present Avenanthramide 2p as a potential chemopreventive dietary compound that merits further study using in vivo models of cancers; they also provide a new perspective on the mechanism of action of Triptolide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5682
FH535, ≥98% (HPLC)
C13H10Cl2N2O4S