EMAIL THIS PAGE TO A FRIEND
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences
[Progress in determination of histamine levels in biological samples].
PMID 23239662
Abstract
Neuronal histamine is crucially involved in a number of physiological functions as well as in neuropsychiatric diseases. Determination of histamine in biological samples is thus of importance in the clinical studies. The aim of this review is to summarize the progress or effort made in this field, with focus on the high-performance liquid chromatography.