EMAIL THIS PAGE TO A FRIEND
Blood
Sense and nonsense of high-dose cytarabine for acute myeloid leukemia.
PMID 23287624
Abstract
High-dose cytarabine applied during remission induction or as consolidation after attainment of a complete remission has become an established element in the treatment of adults with acute myeloid leukemia. Recent evidence has challenged the need for these exceptionally high-dose levels of cytarabine. In this review, we present a reappraisal of the usefulness of high-dose cytarabine for acute myeloid leukemia treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1768
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
C9H13N3O5