EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Potent oral anticonvulsant action of CPP and CPPene in DBA/2 mice.
PMID 2332032
Abstract
CPP (3-(2-carboxypiperazine-4-yl)-1-phosphonate), and its unsaturated analogue, CPPene (3-(2-carboxypiperazine-4-yl)-1-propenyl-1-phosphonic acid), have potent anticonvulsant activity against sound-induced clonic seizures in DBA/2 mice. Following i.p. administration the protection is maximal at 1-2 h, and the ED50 values (mumol/kg) are: D(-)-CPPene, 1.54; D(-)-CPP, 2.75; D,L( +/- )-CPP, 4.36. Following oral administration the protection is maximal at 3-4 h and the ED50 (mumol/kg) values are: D(-)-CPPene, 40.19; D(-)-CPP, 65.80; D,L( +/- )-CPP, 108.1.