EMAIL THIS PAGE TO A FRIEND
Cancer letters
Gastrin acting on the cholecystokinin2 receptor induces cyclooxygenase-2 expression through JAK2/STAT3/PI3K/Akt pathway in human gastric cancer cells.
PMID 23376640
Abstract
Gastrin, cholecystokinin2 receptor (CCK2R), and cyclooxygenase-2 (COX-2) have been implicated in the carcinogenesis and progression of gastric cancer. Our study demonstrated that antagonist or siRNA against CCK2R blocked amidated gastrin (G17)-induced activation of STAT3 and Akt in gastric cancer cell lines. G17-increased COX-2 expression and cell proliferation were effectively blocked by CCK2R antagonist and inhibitors of JAK2 and PI3K. In addition, knockdown of STAT3 expression significantly attenuated G17-induced PI3K/Akt activation, COX-2 expression, and cell proliferation. These results suggest that CCK2R-mediated COX-2 up-regulation via JAK2/STAT3/PI3K/Akt pathway is involved in the proliferative effect of G17 on human gastric cancer cells.