EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Montmorillonite K-10 catalyzed green synthesis of 2,6-unsubstituted dihydropyridines as potential inhibitors of PDE4.
PMID 23380174
Abstract
Montmorillonite K-10 mediated MCR of anilines, arylaldehydes and ethyl-3,3-diethoxypropionate in water afforded 2,6-unsubstituted dihydropyridines depending on the nature of anilines employed. A variety of dihydropyridines were prepared by using this green methodology in good yields and montmorillonite K-10 was found to be an inexpensive and reusable catalyst. The structure elaboration of a representative compound was carried out under Heck conditions. Some of the compounds synthesized showed significant inhibition of PDE4B when tested in vitro. Docking studies indicated that one of the ester moieties of these compounds played a key role in their interactions with the PDE4B protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414476
Ethyl 3,3-diethoxypropionate, 90%, technical grade
C9H18O4