PloS one
Stat3 inhibitor Stattic exhibits potent antitumor activity and induces chemo- and radio-sensitivity in nasopharyngeal carcinoma.
PMID 23382914
Abstract
Nasopharyngeal carcinoma (NPC) is an Epstein-Barr virus-associated malignancy most common in East Asia, Africa and Alaska. Radiotherapy and cisplatin-based chemotherapy are the main treatment options. Unfortunately, disease response to concurrent chemoradiotherapy varies among patients with NPC, and many cases are resistant to cisplatin and radiotherapy. Signal transducer and activator of transcription 3 (Stat3) has been implicated in the development and progression of various solid tumors. In this study, we assessed the activation and expression of Stat3 in NPC cells. We found that Stat3 was activated and could be blocked by the small molecule inhibitor Stattic. The inhibition of Stat3 in NPC cells by Stattic decreased the expression of cyclin D1 in a dose- and time-dependent manner. Thus, Stattic was used to target Stat3 in NPC cell lines. We found that Stattic could inhibit cell viability and proliferation in NPC cells and significantly induced apoptosis. Additionally, Stat3 transfection attenuated, whereas Stat3 knockdown enhanced, the effects of Stattic upon cell viability inhibition and apoptosis induction. Furthermore, Stattic sensitized NPC cells to cisplatin and ionizing radiation (IR) by preventing cell proliferation and inducing apoptosis. Taken together, Stattic inhibit Stat3 and display antitumor effect in NPC, and enhanced chemosensitivity and radiosensitivity in NPC. Therefore, our findings provide the base for more rational approaches to treat NPC in the clinic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7947
Stattic, ≥98% (HPLC), powder
C8H5NO4S