PloS one
The brominated compound aeroplysinin-1 inhibits proliferation and the expression of key pro- inflammatory molecules in human endothelial and monocyte cells.
PMID 23383109
Abstract
Aeroplysinin-1 is a brominated antibiotic used by some sponges for defense against bacterial pathogen invasion. Aeroplysinin-1 has a wide spectrum of anti-tumoral action and behaves as a potent anti-angiogenic compound for bovine aortic endothelial cells. In this study, we demonstrate anti-angiogenic effects of aeroplysinin-1 on human endothelial cells. Furthermore, the response of angiogenesis related genes to aeroplysinin-1 treatment was studied in human endothelial cells by using gene arrays. The major changes were observed in thrombospondin 1 (TSP-1) and monocyte chemoattractant protein-1 (MCP-1), both of which were down-regulated. These inhibitory effects of aeroplysinin-1 were confirmed by using independent experimental approaches. To have a deeper insight on the anti-inflammatory effects of aeroplysinin-1 in endothelial cells, cytokine arrays were also used. This experimental approach confirmed effects on MCP-1 and TSP-1 and showed down-regulation of several other cytokines. Western blotting experiments confirmed down-regulation of ELTD1 (EGF, latrophilin and seven transmembrane domain-containing protein 1), interleukin 1α and matrix metalloproteinase 1 (MMP-1). These results along with our observation of a dramatic inhibitory effect of aeroplysinin-1 on cyclooxygenase-2 protein expression levels in endothelial cells and a human monocyte cell line suggest that aeroplysinin-1 could be a novel anti-inflammatory compound with potential pharmacological interest.