EMAIL THIS PAGE TO A FRIEND
Experimental and clinical transplantation : official journal of the Middle East Society for Organ Transplantation
Conversion of calcineurin inhibitors with mammalian target of rapamycin inhibitors after kidney transplant.
PMID 23387537
Abstract
One way to overcome chronic allograft nephropathy induced by calcineurin inhibitors in immunosuppression protocols for organ transplants is to replace such inhibitors with mammalian target of rapamycin inhibitors, which are not clinically nephrotoxic because they have better renal function. If patients tolerate replacement, there could be a clear preference for mammalian target of rapamycin inhibitors as a maintenance immunosuppressant after renal transplant. This replacement could be sufficient if it were used for a certain time after calcineurin inhibitors. This review considers the conversion effects of calcineurin inhibitors with mammalian target of rapamycin inhibitors from the view point of kidney function during different periods after a kidney transplant.