EMAIL THIS PAGE TO A FRIEND
Optics express
Plasmon hybridization for enhanced nonlinear optical response.
PMID 23388818
Abstract
We report the plasmon hybridization between silver nanoprisms and a thin gold film as a means to tune the plasmon resonance and to achieve enhanced optical second harmonic generation. The hybridization enhances the second harmonic counts by nearly three orders of magnitude when varying the spacer layer between the nanoprisms and the gold film. Finite-difference time-domain calculations agree within a factor of 2 with the experimental findings in terms of the predicted enhancement factor. This plasmon hybridization approach is promising for future applications, including multi-photon lithography and nonlinear sensing using metal nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
296295
Anisole, anhydrous, 99.7%
C7H8O
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3