EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Taurine and its neuroprotective role.
PMID 23392921
Abstract
Taurine plays multiple roles in the CNS including acting as a -neuro-modulator, an osmoregulator, a regulator of cytoplasmic calcium levels, a trophic factor in development, and a neuroprotectant. In neurons taurine has been shown to prevent mitochondrial dysfunction and to protect against endoplasmic reticulum (ER) stress associated with neurological disorders. In cortical neurons in culture taurine protects against excitotoxicity through reversing an increase in levels of key ER signaling components including eIF-2-alpha and cleaved ATF6. The role of communication between the ER and mitochondrion is also important and examples are presented of protection by taurine against ER stress together with prevention of subsequent mitochondrial initiated apoptosis.