EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Chronic leucine supplementation increases body weight and insulin sensitivity in rats on high-fat diet likely by promoting insulin signaling in insulin-target tissues.
PMID 23404947
Abstract
This study investigated the effect of chronic leucine supplementation on insulin sensitivity and the associated mechanisms in rats on high-fat diet (HFD). Male Sprague-Dawley rats were fed either normal chow diet or HFD supplemented with 0, 1.5, 3.0, and 4.5% leucine for 24xa0weeks. We found that chronic leucine supplementation increased insulin sensitivity together with increased body weight in rats on HFD, but had no effect on insulin sensitivity in rats on normal chow diet. The increased insulin sensitivity by leucine supplementation was not associated with altered ectopic fat accumulation in liver and muscle, plasma levels of lipids and cytokines, but is associated with reduced oxidative stress and improved insulin signaling. Chronic leucine supplementation did not enhance insulin receptor substract-1 (IRS-1) phosphorylation on serine 302, but elevated basal IRS-1 phosphorylation on tyrosine 632 and improved insulin-stimulated protein kinase B (Akt) and mammalian target of rapamycin (mTOR) phosphorylation in liver, skeletal muscle, and adipose tissue of rats on HFD rats, indicating leucine supplementation prevented HFD-induced insulin resistance in insulin-target tissues. Chronic leucine supplementation can increase insulin sensitivity and body weight likely by reducing oxidative stress and improving insulin signaling pathway in rats on HFD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0878
O-Phospho-L-serine
C3H8NO6P