EMAIL THIS PAGE TO A FRIEND
Avicenna journal of medical biotechnology
Production and purification of streptokinase by protected affinity chromatography.
PMID 23407807
Abstract
Streptokinase is an extracellular protein, extracted from certain strains of beta hemolytic streptococcus. It is a non-protease plasminogen activator that activates plasminogen to plasmin, the enzyme that degrades fibrin cloth through its specific lysine binding site; it is used therefore as a drug in thrombolytic therapy. The rate of bacterial growth and streptokinase production was studied in condition of excess glucose addition to culture media and its pH maintenance. The streptokinase product of the bacterial culture was preliminary extracted by salt precipitation and then purified by affinity chromatography on plasminogen substituted sepharose-4B in a condition that the plasminogen active site was protected from streptokinase-induced activation. The purity of streptokinase was confirmed by SDS-PAGE and its biological activity determined in a specific streptokinase assay. The results showed that in the fed-batch culture, the rate of streptokinase production increased over two times as compared with the batch culture while at the same time, shortening the streptokinase purification to a single step increased the yield over 95% at the chromatography stage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8010
4-Nitrophenyl 4-guanidinobenzoate hydrochloride, protease inhibitor and substrate
C14H12N4O4 · HCl