EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Rig-G negatively regulates SCF-E3 ligase activities by disrupting the assembly of COP9 signalosome complex.
PMID 23415865
Abstract
We previously showed that Rig-G, an antiproliferative protein induced by interferon, can sequester CSN5 protein in the cytoplasm. Here, we report that Rig-G can destroy the functions of CSN5-containing COP9 signalosome (CSN), a highly conserved multiprotein complex implicated in protein deneddylation, deubiquitination, and phosphorylation. By damaging integrity and stability of the CSN complex, Rig-G can dramatically reduce the cellular content of CSN complex and inhibit its regulatory roles in assembly and activation of cullin-RING ubiquitin E3 ligases (CRL). Furthermore, Rig-G can cause excessive activation of CRL through inhibition of CSN-mediated deneddylation, largely decreasing protein levels of Cul1 and βTrCP, two important subunits of SCF (Skp1-Cul1-F-box protein)-E3 ligase. Rig-G can also attenuate the ability of CSN to recruit USP15 and impair CSN-associated deubiquitination. Increased autoubiquitination of βTrCP and concomitant accumulation of target substrates (such as IκBα) are observed in Rig-G-expressing cells. Taken together, our findings reveal for the first time the negative regulation of Rig-G on SCF-E3 ligase activities through disrupting CSN complex, not only contributing to further investigation on biological functions of Rig-G, but also leading to better understanding of the CSN complex as a potential target in tumor diagnosis and treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10468 CompoZr® Knockout ZFN Kit, Human IFIT3 (NM_001031683)