EMAIL THIS PAGE TO A FRIEND
Oncology letters
Serum response factor is overexpressed in esophageal squamous cell carcinoma and promotes Eca-109 cell proliferation and invasion.
PMID 23426188
Abstract
Recent studies indicate that serum response factor (SRF) is highly expressed in tumors such as hepatocellular, thyroid, esophageal and lung carcinoma. However, the expression and roles of SRF in esophageal squamous cell carcinoma (ESCC) are unclear. In this study, immunohistochemistry was used to compare the expression of SRF in ESCC cases (n=73) and normal controls (n=30). The RNA interference (RNAi) technique was used to knock down the expression of SRF in Eca-109 cells. Cell proliferation, cell cycle stage and invasion were measured with cell counting kit (CCK)-8, flow cytometry and Transwell assays, respectively. Western blotting was used to measure SRF, E-cadherin, β-catenin and cyclin D1 expression in Eca-109 cells treated with siRNA. The study demonstrated that human ESCC has increased expression of SRF. In addition, blocking SRF expression inhibited tumor proliferation and invasion. In conclusion, SRF has the potential to be a new marker for ESCC diagnosis and therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU003471 MISSION® esiRNA, esiRNA targeting human SRF (esiRNA1)