EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Therapeutic efficacy of erythropoietin in experimental autoimmune encephalomyelitis in mice, a model of multiple sclerosis.
PMID 23456868
Abstract
Erythropoietin (EPO) has neuroprotective effects in many models of damage and disease of the nervous system where neuroinflammation plays a substantial role, including experimental autoimmune encephalomyelitis (EAE), the animal model of multiple sclerosis (MS). Since the first pioneering studies, in which EPO was shown to protect rats with acute EAE mainly by inhibiting inflammation, many other studies have pointed out other mechanisms of protection, including oligodendrogenesis and inhibition of axonal damage.Here we review the preclinical studies in which EPO has shown therapeutic efficacy in several models of EAE in mice and rats. Moreover, we report in detail the protocol to administer EPO to mice with myelin oligodendrocyte glycoprotein (MOG)-induced chronic progressive EAE, and a representative result. In this model, EPO inihibits the clinical score of the disease when administered according to a preventive but also to a therapeutic schedule, and therefore at disease onset, suggesting that it might not only inhibit inflammation but also actively stimulate repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1515000 Erythropoietin, BRP, European Pharmacopoeia (EP) Reference Standard