EMAIL THIS PAGE TO A FRIEND
Otology & neurotology : official publication of the American Otological Society, American Neurotology Society [and] European Academy of Otology and Neurotology
Characterization of neuronal cell death in the spiral ganglia of a mouse model of endolymphatic hydrops.
PMID 23462289
Abstract
Spiral ganglion neurons (SGN) in the Phex male mouse, a murine model of postnatal endolymphatic hydrops (ELH) undergo progressive deterioration reminiscent of human and other animal models of ELH with features suggesting apoptosis as an important mechanism. Histologic analysis of the mutant's cochlea demonstrates ELH by postnatal Day (P) 21 and SGN loss by P90. The SGN loss seems to occur in a consistent topographic pattern beginning at the cochlear apex. SGN were counted at P60, P90, and P120. Semiquantitative reverse transcriptase-polymerase chain reaction (RT-PCR), quantitative PCR, and immunohistochemical analyses of activated caspase-3, caspase-8, and caspase-9 were performed on cochlear sections obtained from mutants and controls. Terminal deoxynucleotidyl transferase-mediated dUTP-biotin nick-end labeling assay (TUNEL) was carried out on 2 mutants and 2 controls. Corrected SGN counts in control mice were greater in the apical turn of the cochleae at P90 and P120, respectively (p < 0.01). Increased expression of activated caspase-3, caspase-8, and caspase-9 was seen in the mutant. At later time points, activated caspase expression gradually declined in the apical turns and increased in basal turns of the cochlea. Quantitative and semiquantitative PCR analysis confirmed increased expression of caspase-3, caspase-8, and caspase-9 at P21 and P40. TUNEL staining demonstrated apoptosis at P90 in the apical and basal turns of the mutant cochleae. SGN degeneration in the Phex /Y mouse seems to mimic patterns observed in other animals with ELH. Apoptosis plays an important role in the degeneration of the SGN in the Phex male mouse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11684795910
In Situ Cell Death Detection Kit, Fluorescein