EMAIL THIS PAGE TO A FRIEND
Journal of the renin-angiotensin-aldosterone system : JRAAS
Impact of taurine supplementation on blood pressure in gestational protein-restricted offspring: Effect on the medial solitary tract nucleus cell numbers, angiotensin receptors, and renal sodium handling.
PMID 23468165
Abstract
The current study considers changes of the postnatal brainstem cell number and angiotensin receptors by maternal protein restriction (LP) and LP taurine supplementation (LPT), and its impact on arterial hypertension development in adult life. The brain tissue studies were performed by immunoblotting, immunohistochemistry, and isotropic fractionator analysis. The current study shows that elevated blood pressure associated with decreased fractional urinary sodium excretion (FENa) in adult LP offspring was reverted by diet taurine supplementation. Also, that 12-day-old LP pups present a reduction of 21% of brainstem neuron counts, and, immunohistochemistry demonstrates a decreased expression of type 1 angiotensin II receptors (AT1R) in the entire medial solitary tract nuclei (nTS) of 16-week-old LP rats compared to age-matched NP and LPT offspring. Conversely, the immunostained type 2 AngII (AT2R) receptors in 16-week-old LP nTS were unchanged. The present investigation shows a decreased FENa that occurs despite unchanged creatinine clearance. It is plausible to hypothesize an association of decreased postnatal nTS cell number, AT1R/AT2R ratio and FENa with the higher blood pressure levels found in taurine-deficient progeny (LP) compared with age-matched NP and LPT offspring.