EMAIL THIS PAGE TO A FRIEND
Ying yong sheng tai xue bao = The journal of applied ecology
[Migration and transformation of anthropogenic platinum group elements in environment: a review].
PMID 23479898
Abstract
Anthropogenic platinum group elements (PGEs) are widely applied in vehicle exhaust catalytic converters (VECs), industrial catalysts, and pharmaceutics, making the PGEs, especially Pt, Pd, and Rh, become the newly environmental pollutants in some fields. Given the positive correlations between the Pt/Pd and Pt/Rh ratios in various environmental samples and the active components of VECs, the VECs containing PGEs as catalysts are regarded as the primary source of PGEs pollution. Sufficient reports indicated that in the past three decades, there was a significant increase of PGEs concentrations in diverse environmental matrices like airborne particulate matters, aquatic ecosystem components (e.g., river water, rain water, groundwater, seawater, and sediments), soils, road dusts, and organisms. It was generally assumed that anthropogenic PGEs behave in inert manner, and the health risks associated with the environmental exposures to PGEs are minimal. However, the recent studies on PGEs toxicity and environmental bioavailability indicated that once entering environment, anthropogenic PGEs might easily be mobilized and transformed into more toxic forms under the actions of various biogeochemical processes, and thereby, enhanced their bioavailability and posed potential health risks to human beings through food chain. This paper summarized the research results about the sources, distribution, and biogeochemical behaviors of PGEs in various environmental media, and it was considered that to establish the standards of PGEs for human health risks, to develop standard substances of PGEs for environmental measurements, to study the PGEs in the sediments of marginal seas, and to assess the toxicity of PGEs to marine mollusks, the present contamination status of PGEs in foods, and the risks of PGEs to human health would be the hot research topics in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373214
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Pt
373222
Platinum, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Pt
373230
Platinum, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Pt
349372
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Pt
267260
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Pt
305189
Platinum, foil, thickness 0.5 mm, 99.9% trace metals basis
Pt
349321
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Pt
327425
Platinum, foil, thickness 0.25 mm, 99.9% trace metals basis
Pt
349348
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Pt
327433
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Pt
267252
Platinum, foil, thickness 0.1 mm, 99.99% trace metals basis
Pt
349380
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Pt
349356
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Pt
303798
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Pt
349364
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Pt
267244
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Pt
298107
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Pt
298093
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Pt
685453
Platinum, nanopowder, <50 nm particle size (TEM)
Pt
204048
Platinum, powder (coarse), 99.99% trace metals basis
Pt
204013
Platinum, powder, 99.995% trace metals basis
Pt
327476
Platinum, powder, ≤10 μm, 99.9% trace metals basis
Pt
327484
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Pt
267155
Platinum, sponge, ≥99.9% trace metals basis
Pt
444685
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Pt
349399
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Pt
267236
Platinum, wire, diam. 1.0 mm, 99.99% trace metals basis
Pt
327492
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Pt
267201
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Pt
267228
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Pt