EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Preparation and X-ray structural study of 1-arylbenziodoxolones.
PMID 23480389
Abstract
Various 1-arylbenziodoxolones can be conveniently prepared from 2-iodobenzoic acid and arenes by a one-pot procedure using Oxone (2KHSO5·KHSO4·K2SO4) as an inexpensive and environmentally safe oxidant. This procedure is also applicable for the synthesis of the 7-methylbenziodoxolone ring system using 2-iodo-3-methylbenzoic acid as starting compound. Structures of four 1-arylbenziodoxolone derivatives were established by single-crystal X-ray diffraction analysis. An enhanced reactivity of 1-aryl-7-methylbenziodoxolones toward nucleophiles compared to unsubstituted 1-arylbenziodoxolones has been found.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
418463
2-Iodobenzoic acid, ≥99%, purified by sublimation
C7H5IO2