EMAIL THIS PAGE TO A FRIEND
Organic Letters
First asymmetric total syntheses and determination of absolute configurations of (+)-eudesmadiene-12,6-olide and (+)-frullanolide.
PMID 23484913
Abstract
The first asymmetric total syntheses of sesquiterpene lactones (+)-eudesmadiene-12,6-olide (1) and (+)-frullanolide (2) have been accomplished from 4-bromo-2-methoxyphenol (5) in 12 and 13 synthetic steps, respectively, and the absolute configurations of these two natural products were determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533637
4-Bromo-2-methoxyphenol, 98%
C7H7BrO2
Set your institution to read full text papers when available.