EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Nobiletin attenuates metastasis via both ERK and PI3K/Akt pathways in HGF-treated liver cancer HepG2 cells.
PMID 23537747
Abstract
Hepatocyte growth factor (HGF), and its receptor, c-Met activation has recently been shown to play important roles in cancer invasion and metastasis in a wide variety of tumor cells. We use HGF as an invasive inducer of human HepG2 cells to investigate the effect of four flavones including apigenin, tricetin, tangeretin, and nobiletin on HGF/c-Met-mediated tumor invasion and metastasis. Among them, nobiletin markedly inhibited HGF-induced the abilities of the adhesion, invasion, and migration by cell-matrix adhesion assay and transwell-chamber invasion/migration assay under non-cytotoxic concentrations. Data also showed nobiletin inhibited HGF-induced cell scattering and cytoskeleton changed such as filopodia and lamellipodia. Furthermore, nobiletin could inhibit HGF-induced the membrane localization of phosphorylated c-Met, ERK2, and Akt, but not phosphorylated JNK1/2 and p38. Next, nobiletin significantly decreased the levels of phospho-ERK2 and phospho-Akt in ERK2 or Akt siRNA-transfected cells concomitantly with a marked reduction on cell invasion and migration. In conclusion, nobiletin attenuates HGF-induced HepG2 cells metastasis involving both ERK and PI3K/Akt pathways and are potentially useful as anti-metastatic agents for the treatment of hepatoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
42251
Apigenin, analytical standard
C15H10O5
01760595
Apigenin, primary pharmaceutical reference standard
C15H10O5
A3145
Apigenin, ≥97% (TLC), from parsley, powder
C15H10O5
10798
Apigenin, ≥95.0% (HPLC)
C15H10O5
D2510
Deoxycholic acid, ≥98% (HPLC)
C24H40O4
D4297
Deoxycholic acid, BioXtra, ≥98% (HPLC)
C24H40O4
30960
Deoxycholic acid, ≥99.0% (T)
C24H40O4
EHU083501
MISSION® esiRNA, esiRNA human AKT1 (esiRNA1)
EMU015011 MISSION® esiRNA, esiRNA targeting mouse Akt1 (esiRNA1)
EHU044171 MISSION® esiRNA, esiRNA human MAPK1 (esiRNA1)
N1538
Nobiletin, ≥97%
C21H22O8
92600
Nobiletin, analytical standard
C21H22O8