EMAIL THIS PAGE TO A FRIEND
Endocrine research
Association of TRPC1 gene polymorphisms with type 2 diabetes and diabetic nephropathy in Han Chinese population.
PMID 23544998
Abstract
The recent genome-wide association studies reveal that chromosome 3q resides within the linkage region for diabetic nephropathy (DN) in type 1 and type 2 diabetes mellitus (T1D and T2D). The TRPC1 gene is on chromosome 3q22-24, and it has been demonstrated that TRPC1 expression is reduced in the kidney of diabetic animal models. Genetic association of TRPC1 polymorphism with T1D and DN has been reported in European Americans. However, there are no studies reporting the association of TRPC1 genetic polymorphism with T2D with and without DN in Chinese population. This study aimed to demonstrate the genetic role of TRPC1 in the development of T2D with and without DN in Chinese Han population. A genetic association study of TRPC1 was performed in T2D cases and in nondiabetic controls from Han population located in Northern Chinese areas. Six tag single nucleotide polymorphism (SNP) markers derived from HapMap data were genotyped. Among the six SNPs, only rs7638459 was suspected as risk factor of T2D without DN, fitting the log-additive model. The adjusted odds ratio (OR) for the CC genotyping was 2.39 (95% confidence interval (CI) = 1.00-5.68), compared with the TT genotyping. In addition, rs953239 was found to be a protective factor of getting DN in T2D, also fitting the log-additive model. When compared with the AA genotyping for SNP rs953239, the adjusted OR for CC genotyping was 0.63 (95% CI = 0.44-0.99). To summarize, this study shows that TRPC1 genetic polymorphisms are associated with T2D and DN in T2D in the Han Chinese population.