EMAIL THIS PAGE TO A FRIEND
Journal of tissue engineering and regenerative medicine
Agonism of Wnt-β-catenin signalling promotes mesenchymal stem cell (MSC) expansion.
PMID 23554411
Abstract
Promoting mesenchymal stem cell (MSC) proliferation has numerous applications in stem cell therapies, particularly in the area of regenerative medicine. In order for cell-based regenerative approaches to be realized, MSC proliferation must be achieved in a controlled manner without compromising stem cell differentiation capacities. Here we demonstrate that 6-bromoindirubin-3'-oxime (BIO) increases MSC β-catenin activity 106-fold and stem cell-associated gene expression ~33-fold, respectively, over untreated controls. Subsequently, BIO treatment increases MSC populations 1.8-fold in typical 2D culture conditions, as well as 1.3-fold when encapsulated within hydrogels compared to untreated cells. Furthermore, we demonstrate that BIO treatment does not reduce MSC multipotency where MSCs maintain their ability to differentiate into osteoblasts, chondrocytes and adipocytes using standard conditions. Taken together, our results demonstrate BIO's potential utility as a proliferative agent for cell transplantation and tissue regeneration.