EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
Ugi/Robinson-Gabriel reactions directed toward the synthesis of 2,4,5-trisubstituted oxazoles.
PMID 23559684
Abstract
This Letter discloses a novel concise synthesis of a series of 2,4,5-trisubstituted oxazoles via a tandem Ugi/Robinson-Gabriel sequence. Herein, 2,4-dimethoxybenzylamine 1 was used as an ammonia equivalent in combination with arylglyoxal 3 and supporting Ugi reagents, an isonitrile and carboxylic acid. As such the product of the acid treated Ugi intermediate is ideally configured to undergo a Robinson-Gabriel cyclodehydration reaction to yield the desired oxazole scaffold 5.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
432725
2,4-Dimethoxybenzylamine, 98%
C9H13NO2