EMAIL THIS PAGE TO A FRIEND
Acta neuropathologica
Meningeal hemangiopericytoma and solitary fibrous tumors carry the NAB2-STAT6 fusion and can be diagnosed by nuclear expression of STAT6 protein.
PMID 23575898
Abstract
Non-central nervous system hemangiopericytoma (HPC) and solitary fibrous tumor (SFT) are considered by pathologists as two variants of a single tumor entity now subsumed under the entity SFT. Recent detection of frequent NAB2-STAT6 fusions in both, HPC and SFT, provided additional support for this view. On the other hand, current neuropathological practice still distinguishes between HPC and SFT. The present study set out to identify genes involved in the formation of meningeal HPC. We performed exome sequencing and detected the NAB2-STAT6 fusion in DNA of 8/10 meningeal HPC thereby providing evidence of close relationship of these tumors with peripheral SFT. Due to the considerable effort required for exome sequencing, we sought to explore surrogate markers for the NAB2-STAT6 fusion protein. We adopted the Duolink proximity ligation assay and demonstrated the presence of NAB2-STAT6 fusion protein in 17/17 HPC and the absence in 15/15 meningiomas. More practical, presence of the NAB2-STAT6 fusion protein resulted in a strong nuclear signal in STAT6 immunohistochemistry. The nuclear reallocation of STAT6 was detected in 35/37 meningeal HPC and 25/25 meningeal SFT but not in 87 meningiomas representing the most important differential diagnosis. Tissues not harboring the NAB2-STAT6 fusion protein presented with nuclear expression of NAB2 and cytoplasmic expression of STAT6 proteins. In conclusion, we provide strong evidence for meningeal HPC and SFT to constitute variants of a single entity which is defined by NAB2-STAT6 fusion. In addition, we demonstrate that this fusion can be rapidly detected by STAT6 immunohistochemistry which shows a consistent nuclear reallocation. This immunohistochemical assay may prove valuable for the differentiation of HPC and SFT from other mesenchymal neoplasms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DUO92012
Duolink® In Situ Detection Reagents Brightfield
DUO92013
Duolink® In Situ Detection Reagents FarRed
DUO92014
Duolink® In Situ Detection Reagents Green
DUO92007
Duolink® In Situ Detection Reagents Orange
DUO92008
Duolink® In Situ Detection Reagents Red
DUO92106
Duolink® In Situ Orange Starter Kit Goat/Rabbit
DUO92104
Duolink® In Situ Orange Starter Kit Mouse/Goat
DUO92102
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
DUO92006
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92003
Duolink® In Situ PLA® Probe Anti-Goat PLUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92004
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92001
Duolink® In Situ PLA® Probe Anti-Mouse PLUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92005
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92002
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92010 Duolink® In Situ Probemaker MINUS
DUO92009 Duolink® In Situ Probemaker PLUS
DUO92105
Duolink® In Situ Red Starter Kit Goat/Rabbit
DUO92103
Duolink® In Situ Red Starter Kit Mouse/Goat
DUO92101
Duolink® In Situ Red Starter Kit Mouse/Rabbit