EMAIL THIS PAGE TO A FRIEND
Environmental science. Processes & impacts
The impact of particle size selective sampling methods on occupational assessment of airborne beryllium particulates.
PMID 23575953
Abstract
In 2010, the American Conference of Governmental Industrial Hygienists (ACGIH) formally changed its Threshold Limit Value (TLV) for beryllium from a 'total' particulate sample to an inhalable particulate sample. This change may have important implications for workplace air sampling of beryllium. A history of particle size-selective sampling methods, with a special focus on beryllium, will be provided. The current state of the science on inhalable sampling will also be presented, including a look to the future at what new methods or technology may be on the horizon. This includes new sampling criteria focused on particle deposition in the lung, proposed changes to the existing inhalable convention, as well as how the issues facing beryllium sampling may help drive other changes in sampling technology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF12366660
Beryllium, rod, 100mm, diameter 7.0mm, 99%
Be
GF41430787
Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%
Be
GF53985370
Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%
Be
GF46045627
Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%
Be
GF95797692
Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%
Be
GF09275823
Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%
Be
GF73403142
Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%
Be
GF58877948
Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%
Be
GF45831236
Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%
Be
GF96781214
Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%
Be
GF60949840
Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%
Be
GF59478586
Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%
Be
GF75393388
Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%
Be
GF59743738
Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%
Be
GF29665086
Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%
Be
GF98865129
Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%
Be
GF79324959
Beryllium, wire reel, 0.2m, diameter 0.85mm, annealed and clean, 99%
Be
GF67966385
Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%
Be
GF11051558
Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%
Be
GF34823224
Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%
Be
GF24172703
Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%
Be
GF81444764
Beryllium, wire reel, 1m, diameter 0.05mm, annealed and clean, 99.7%
Be
GF52902840
Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%
Be
GF21588565
Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%
Be
GF80595793
Beryllium, wire reel, 5m, diameter 0.05mm, annealed and clean, 99.7%
Be
GF02814702
Beryllium, mesh, 5x5mm, nominal aperture 0.13mm, thickness 0.025mm, wire diameter 0.1mm, 100 wires/inch, open area 50%, electro-formed mesh
Be
GF06987224
Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%
Be
GF21271025
Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%
Be
GF28341840
Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%
Be
GF65661363
Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%
Be