EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Quercetin suppressed CYP2E1-dependent ethanol hepatotoxicity via depleting heme pool and releasing CO.
PMID 23583009
Abstract
Naturally occuring quercetin protects hepatocytes from ethanol-induced oxidative stress, and heme oxygenase-1 (HO-1) induction and carbon monoxide (CO) metabolite may be implicated in the beneficial effect. However, the precise mechanism by which quercetin counteracts CYP2E1-mediated ethanol hepatotoxicity through HO-1 system is still remained unclear. To explore the potential mechanism, herein, ethanol (4.0 g/kg.bw.) was administrated to rats for 90 days. Our data showed that chronic ethanol over-activated CYP2E1 but suppressed HO-1 with concurrent hepatic oxidative damage, which was partially normalized by quercetin (100mg/kg.bw.). Quercetin (100 μM) induced HO-1 and depleted heme pool when incubated to human hepatocytes. Ethanol-stimulated (100mM) CYP2E1 upregulation was suppressed by quercetin but further enhanced by HO-1 inhibition with resultant heme accumulation. CO scavenging blocked the suppression of quercetin only on CYP2E1 activity. CO donor dose-dependently inactivated CYP2E1 of ethanol-incubated microsome, which was mimicked by HO-1 substrate but abolished by CO scavenger. Thus, CYP2E1-mediated ethanol hepatotoxicity was alleviated by quercetin through HO-1 induction. Depleted heme pool and CO releasing limited protein synthesis and inhibited enzymatic activity of CYP2E1, respectively.