PloS one
p38 MAPK regulates cavitation and tight junction function in the mouse blastocyst.
PMID 23593143
Abstract
Blastocyst formation is essential for implantation and maintenance of pregnancy and is dependent on the expression and coordinated function of a series of proteins involved in establishing and maintaining the trans-trophectoderm ion gradient that enables blastocyst expansion. These consist of Na/K-ATPase, adherens junctions, tight junctions (TJ) and aquaporins (AQP). While their role in supporting blastocyst formation is established, the intracellular signaling pathways that coordinate their function is unclear. The p38 MAPK pathway plays a role in regulating these proteins in other cell types and is required for embryo development at the 8-16 cell stage, but its role has not been investigated in the blastocyst. p38 MAPK regulates blastocyst formation by regulating blastocyst formation gene expression and function. Embryos were cultured from the early blastocyst stage for 12 h or 24 h in the presence of a potent and specific p38 MAPK inhibitor, SB 220025. Blastocyst expansion, hatching, gene family expression and localization, TJ function and apoptosis levels were analyzed. Inhibition of the p38 MAPK pathway reduced blastocyst expansion and hatching, increased tight junction permeability, affected TJP1 localization, reduced Aqp3 expression, and induced a significant increase in apoptosis. The p38 MAPK pathway coordinates the overall events that regulate blastocyst formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0679
3,6-Diacetoxyphthalonitrile
C12H8N2O4