EMAIL THIS PAGE TO A FRIEND
Immunologic research
Aluminum in the central nervous system (CNS): toxicity in humans and animals, vaccine adjuvants, and autoimmunity.
PMID 23609067
Abstract
We have examined the neurotoxicity of aluminum in humans and animals under various conditions, following different routes of administration, and provide an overview of the various associated disease states. The literature demonstrates clearly negative impacts of aluminum on the nervous system across the age span. In adults, aluminum exposure can lead to apparently age-related neurological deficits resembling Alzheimer's and has been linked to this disease and to the Guamanian variant, ALS-PDC. Similar outcomes have been found in animal models. In addition, injection of aluminum adjuvants in an attempt to model Gulf War syndrome and associated neurological deficits leads to an ALS phenotype in young male mice. In young children, a highly significant correlation exists between the number of pediatric aluminum-adjuvanted vaccines administered and the rate of autism spectrum disorders. Many of the features of aluminum-induced neurotoxicity may arise, in part, from autoimmune reactions, as part of the ASIA syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
356859
Aluminum, foil, thickness 0.05 mm, 99.8% trace metals basis
Al
11008
Aluminum, grit, ≥97.0% (complexometric)
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
11010
Aluminum, powder (fine), ≥93% (complexometric)
Al
424803
Aluminum, disc, 0.5-1.7 mm, ≥99.99% trace metals basis
Al
266590
Aluminum, foil, thickness 2.0 mm, 99.999%
Al
326909
Aluminum, wire, diam. 0.127 mm, ≥99.99%
Al
IRMM523A
Aluminum, IRMM® certified Reference Material, 0.1 mm foil
Al
IRMM523B
Aluminum, IRMM® certified Reference Material, 1.0 mm foil
Al
IRMM523C
Aluminum, IRMM® certified Reference Material, 1.0 mm wire
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
773964
Aluminum, ingot, L × W 10 mm × 7 mm, 99.99995% trace metals basis
Al
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al