EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Porous polyimide membranes prepared by wet phase inversion for use in low dielectric applications.
PMID 23615465
Abstract
A wet phase inversion process of polyamic acid (PAA) allowed fabrication of a porous membrane of polyimide (PI) with the combination of a low dielectric constant (1.7) and reasonable mechanical properties (Tensile strain: 8.04%, toughness: 3.4 MJ/m3, tensile stress: 39.17 MPa, and young modulus: 1.13 GPa), with further thermal imidization process of PAA. PAA was simply synthesized from purified pyromellitic dianhydride (PMDA) and 4,4-oxydianiline (ODA) in two different reaction solvents such as γ-butyrolactone (GBL) and N-methyl-2-pyrrolidinone (NMP), which produce Mw/PDI of 630,000/1.45 and 280,000/2.0, respectively. The porous PAA membrane was fabricated by the wet phase inversion process based on a solvent/non-solvent system via tailored composition between GBL and NMP. The porosity of PI, indicative of a low electric constant, decreased with increasing concentration of GBL, which was caused by sponge-like formation. However, due to interplay between the low electric constant (structural formation) and the mechanical properties, GBL was employed for further exploration, using toluene and acetone vs. DI-water as a coagulation media. Non-solvents influenced determination of the PAA membrane size and porosity. With this approach, insight into the interplay between dielectric properties and mechanical properties will inform a wide range of potential low-k material applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412287
Pyromellitic dianhydride, 97%
C10H2O6