EMAIL THIS PAGE TO A FRIEND
Odontology
Proteomic analysis of the biological response of MG63 osteoblast-like cells to titanium implants.
PMID 23665890
Abstract
Understanding of the interaction between human MG63 osteoblast-like cells and surfaces is necessary in the field of tissue engineering and biomaterials. Various titanium surfaces are widely used as not only implant materials, but also as miniscrews in orthodontics. Our goal was to assess the proteomic response of MG63 osteoblast-like cells to different titanium surfaces. MG63 osteoblast-like cells were cultured on three different titanium surfaces: a smooth surface (S), a sandblasted with large grit and acid-etched surface (SLA), and a surface coated with a thin layer of hydroxyapatite (HA). Cells grown on the rougher surfaces (SLA and HA) exhibited downregulated cell proliferation and morphological changes. In the proteomic analysis, cells grown on the SLA surface showed upregulated expression of protocadherin-β3 precursor, kinase insert domain receptor, fibroblast growth factor receptor-3, and insulin-like growth factor I, while the expression levels of cell adhesion kinase, collagen α-1(I) chain precursor, collagen type XI α2, and cadherin-11 were upregulated in cells grown on the HA surface. These proteins are known to be involved in osteoblast adhesion, growth, and differentiation. Thus, the surface properties of dental materials can influence the expression of proteins involved in osseointegration-related processes. Proteomic analysis may reveal changes in novel proteins that explain why osseointegration varies depending on surface properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2106696
Anti-BMP2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1411278
Anti-BMP2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA017385
Anti-BMPR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411292
Anti-BMPR2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501745
Anti-CDH11 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501326
Anti-CDH11 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4500033
Anti-CDH11 antibody produced in rabbit, affinity isolated antibody
WH0000650M2 Monoclonal Anti-BMP2 antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution
SAB1403614
Monoclonal Anti-BMPR2 antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB5300504 Monoclonal Anti-BMPR2 antibody produced in mouse, clone 1F12, ascites fluid
SAB5300505 Monoclonal Anti-BMPR2 antibody produced in mouse, clone 3F6, ascites fluid
SAB1402129
Monoclonal Anti-BMPR2 antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
WH0001009M1
Monoclonal Anti-CDH11 antibody produced in mouse, clone 4D10, purified immunoglobulin, buffered aqueous solution