EMAIL THIS PAGE TO A FRIEND
Advances in nutrition (Bethesda, Md.)
Magnesium in disease prevention and overall health.
PMID 23674807
Abstract
Magnesium is the fourth most abundant mineral and the second most abundant intracellular divalent cation and has been recognized as a cofactor for >300 metabolic reactions in the body. Some of the processes in which magnesium is a cofactor include, but are not limited to, protein synthesis, cellular energy production and storage, reproduction, DNA and RNA synthesis, and stabilizing mitochondrial membranes. Magnesium also plays a critical role in nerve transmission, cardiac excitability, neuromuscular conduction, muscular contraction, vasomotor tone, blood pressure, and glucose and insulin metabolism. Because of magnesium's many functions within the body, it plays a major role in disease prevention and overall health. Low levels of magnesium have been associated with a number of chronic diseases including migraine headaches, Alzheimer's disease, cerebrovascular accident (stroke), hypertension, cardiovascular disease, and type 2 diabetes mellitus. Good food sources of magnesium include unrefined (whole) grains, spinach, nuts, legumes, and white potatoes (tubers). This review presents recent research in the areas of magnesium and chronic disease, with the goal of emphasizing magnesium's role in disease prevention and overall health.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF61950618
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF23443646
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF54671924
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF28467636
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF72273157
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF78647293
Magnesium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF43827071
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF30456053
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF33990598
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF58454983
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF34143803
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF30911778
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF78384437
Magnesium, wire reel, 2m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF07006328
Magnesium, wire reel, 2m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF48824208
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF06872924
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF56273777
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg