EMAIL THIS PAGE TO A FRIEND
Cerebral cortex (New York, N.Y. : 1991)
Late maternal hypothyroidism alters the expression of Camk4 in neocortical subplate neurons: a comparison with Nurr1 labeling.
PMID 23680840
Abstract
Maternal thyroid hormones (THs) are essential for normal offspring's neurodevelopment even after onset of fetal thyroid function. This is particularly relevant for preterm children who are deprived of maternal THs following birth, are at risk of suffering hypothyroxinemia, and develop attention-deficit/hyperactivity disorder. Expression of neocortical Ca(2+)/calmodulin kinase IV (Camk4), a genomic target of thyroid hormone, and nuclear receptor-related 1 protein (Nurr1), a postnatal marker of cortical subplate (SP) cells, was studied in euthyroid fetuses and in pups born to dams thyroidectomized in late gestation (LMH group, a model of prematurity), and compared with control and developmentally hypothyroid pups (C and MMI groups, respectively). In LMH pups, the extinction of heavy Camk4 expression in an SP was 1-2 days delayed postnatally compared with C pups. The heavy Camk4 and Nurr1 expression in the SP was prolonged in MMI pups, whereas heavy Camk4 and Nurr1 expression in layer VIb remains at P60. The abnormal expression of Camk4 in the cortical SP and in layer VIb might cause altered cortical connectivity affecting neocortical function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011753
Anti-CAMK4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017206
Anti-CAMK4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37025
Anti-CAMK4 antibody produced in rabbit, IgG fraction of antiserum
SAB4503233
Anti-CAMK4 antibody produced in rabbit, affinity isolated antibody
AV32730
Anti-NR4A2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV38753
Anti-NR4A2 (AB2) antibody produced in rabbit, affinity isolated antibody
N6413
Anti-NR4A2 (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
N6538
Anti-NR4A2 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA000543
Anti-NR4A2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103658
Anti-NR4A2 antibody produced in rabbit, affinity isolated antibody
N4663 Anti-Nurr1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
WH0002571M2
Monoclonal Anti-GAD1 antibody produced in mouse, clone 5E11, purified immunoglobulin, buffered aqueous solution
SAB1403849
Monoclonal Anti-GAD1 antibody produced in mouse, clone 1D12, ascites fluid