EMAIL THIS PAGE TO A FRIEND
BioDrugs : clinical immunotherapeutics, biopharmaceuticals and gene therapy
Everolimus: a guide to its use in liver transplantation.
PMID 23696253
Abstract
The mammalian target of rapamycin inhibitor everolimus (Zortress®, Certican®) was recently approved in the USA and a number of EU countries for use in combination with a reduced dosage of tacrolimus and corticosteroids for the prophylaxis of organ rejection in adult liver transplant recipients. Compared with standard-exposure tacrolimus, early use of everolimus plus a reduced dosage of tacrolimus did not compromise efficacy in liver transplant recipients. In addition, significantly better renal function with everolimus plus reduced-exposure tacrolimus than with standard-exposure tacrolimus was seen from 6 weeks post-transplant onwards. Everolimus plus reduced-exposure tacrolimus has an acceptable tolerability profile in liver transplant recipients.