EMAIL THIS PAGE TO A FRIEND
Disease models & mechanisms
Loss of Pten promotes angiogenesis and enhanced vegfaa expression in zebrafish.
PMID 23720233
Abstract
Angiogenesis, the emergence of vessels from an existing vascular network, is pathologically associated with tumor progression and is of great interest for therapeutic intervention. PTEN is a frequently mutated tumor suppressor and has been linked to the progression of many types of tumors, including hemangiosarcomas in zebrafish. Here, we report that mutant zebrafish embryos lacking functional Pten exhibit enhanced angiogenesis, accompanied by elevated levels of phosphorylated Akt (pAkt). Inhibition of phosphoinositide 3-kinase (PI3K) by LY294002 treatment and application of sunitinib, a widely used anti-angiogenic compound, suppressed enhanced angiogenesis in Pten mutants. Vegfaa has a crucial role in angiogenesis and vegfaa expression was upregulated in embryos lacking functional Pten. Interestingly, vegfaa expression was also upregulated in hemangiosarcomas from haploinsufficient adult zebrafish Pten mutants. Elevated vegfaa expression in mutant embryos lacking functional Pten was suppressed by LY294002. Surprisingly, sunitinib treatment dramatically enhanced vegfaa expression in Pten mutant embryos, which might account for tumor relapse in human patients who are treated with sunitinib. Combined treatment with suboptimal concentrations of sunitinib and LY294002 rescued enhanced angiogenesis in pten mutant embryos without the dramatic increase in vegfaa expression, suggesting a new approach for therapeutic intervention in VEGFR-signaling-dependent tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0012
Sunitinib malate, ≥98% (HPLC)
C22H27FN4O2 · C4H6O5