EMAIL THIS PAGE TO A FRIEND
Revista da Escola de Enfermagem da USP
[Cytotoxicity of PVC tubes sterilized in ethylene oxide after gamma radiation exposure].
PMID 23743920
Abstract
Do materials sterilized using gamma rays become toxic when re-sterilized in ethylene oxide? This question guided the objective of this study, which was to investigate the potential cytotoxic effect of PVC sterilized by gamma radiation and re-sterilized with EO by the agar diffusion method in cell cultures. Nine PVC tubes were subjected to gamma radiation sterilization and were re-sterilized in EO. The tubes were divided into a total of 81 units of analysis that were tested so as to represent the internal and external surfaces and mass of each tube. It was concluded that the PVC materials sterilized in gamma radiation and re-sterilized in EO are not cytotoxic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
387614
Ethylene oxide, ≥99.5%
C2H4O
48838
Ethylene oxide solution, 50 mg/mL in methanol, analytical standard
C2H4O
779202
Ethylene oxide solution, 2.5-3.3 M in THF
C2H4O
81388
Poly(vinyl chloride), low molecular weight
81387
Poly(vinyl chloride), high molecular weight
389293
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
189588
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
389323
Poly(vinyl chloride), average Mw ~80,000, average Mn ~47,000
182621
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
346772
Poly(vinyl chloride)
346764
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
81392
Poly(vinyl chloride), Selectophore, high molecular weight
P2600000
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Set your institution to read full text papers when available.