EMAIL THIS PAGE TO A FRIEND
Clinical and experimental medicine
Immunomodulatory effects of vitamin D in peripheral blood monocyte-derived macrophages from patients with rheumatoid arthritis.
PMID 23824148
Abstract
1,25-Dihydroxyvitamin D (1,25(OH)2D3), the active form of vitamin D, modulates both innate and adaptive immune responses. Emerging epidemiological data has also demonstrated disease-modifying and immunomodulatory effects of vitamin D in a wide range of human autoimmune diseases, including rheumatoid arthritis (RA). To evaluate in vitro effects of 1,25(OH) 2D3 in primary cultures of peripheral blood monocyte-derived macrophages of RA patients, monocyte/macrophages, isolated from peripheral blood mononuclear cells of RA patients and healthy subjects by exploiting their ability to adhere to plastic, were treated with increasing concentrations of 1,25(OH)2D3 for 48 h. TNF-α, IL-1 α, IL-1β, IL-6 and RANKL production was determined by ELISA and nitric oxide (NO) release using the Griess method. Immunocytochemistry analysis was also performed to evaluate alterations in transmembrane TNF-α expression after 1,25(OH) 2D3 treatment. A significant dose-dependent decrease in TNF-α and RANKL production by cultured RA macrophages after 1,25(OH)2D3 treatment was found, whereas a significant reduction in normal cells was observed only at higher concentrations. IL-1 α, IL-1β and IL-6 levels were reduced by 1,25(OH) 2D3 at higher concentrations in all cell populations. TNF-α immunostaining was less intense in treated cells compared with untreated. 1,25(OH) 2D3 significantly reduced NO levels regardless of the concentration used. Vitamin D downregulated proinflammatory mediators in monocyte-derived macrophages, and RA cells appeared more sensitive than normal cells. These effects further provide a rationale for the therapeutic value of vitamin D supplementation in the treatment for RA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C9756
Cholecalciferol, ≥98% (HPLC)
C27H44O
C9774
Cholecalciferol, analytical standard
C27H44O
C1357
Cholecalciferol, meets USP testing specifications
C27H44O
C2100000
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
C27H44O
1131009
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
C27H44O
47763
Cholecalciferol (D3), analytical standard
C27H44O
PHR1237
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
C27H44O
C2150000
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
C27H44O
CKOZFN1756 CompoZr® Knockout ZFN Kit, Human TNF (NM_000594)
CKOZFND1756 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human TNF (NM_000594)
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
740292
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
C27H44O
V-025
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material
C27H44O