EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
TrkB signaling directs the incorporation of newly generated periglomerular cells in the adult olfactory bulb.
PMID 23843518
Abstract
In the adult rodent brain, the olfactory bulb (OB) is continuously supplied with new neurons which survival critically depends on their successful integration into pre-existing networks. Yet, the extracellular signals that determine the selection which neurons will be ultimately incorporated into these circuits are largely unknown. Here, we show that immature neurons express the catalytic form of the brain-derived neurotrophic factor receptor TrkB [full-length TrkB (TrkB-FL)] only after their arrival in the OB, at the time when integration commences. To unravel the role of TrkB signaling in newborn neurons, we conditionally ablated TrkB-FL in mice via Cre expression in adult neural stem and progenitor cells. TrkB-deficient neurons displayed a marked impairment in dendritic arborization and spine growth. By selectively manipulating the signaling pathways initiated by TrkB in vivo, we identified the transducers Shc/PI3K to be required for dendritic growth, whereas the activation of phospholipase C-γ was found to be responsible for spine formation. Furthermore, long-term genetic fate mapping revealed that TrkB deletion severely compromised the survival of new dopaminergic neurons, leading to a substantial reduction in the overall number of adult-generated periglomerular cells (PGCs), but not of granule cells (GCs). Surprisingly, this loss of dopaminergic PGCs was mirrored by a corresponding increase in the number of calretinin+ PGCs, suggesting that distinct subsets of adult-born PGCs may respond differentially to common extracellular signals. Thus, our results identify TrkB signaling to be essential for balancing the incorporation of defined classes of adult-born PGCs and not GCs, reflecting their different mode of integration in the OB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB9042
Anti-Brain Derived Neurotrophic Factor Antibody, pro, 100 µL, Anti-Brain Derived Neurotrophic Factor Antibody, pro detects level of Brain Derived Neurotrophic Factor & has been published & validated for use in IP, WB, IH.
AB1778
Anti-Calbindin D-28K Antibody, 100 µL, This Anti-Calbindin D-28K Antibody is validated for use in IC, IH, IH(P), WB for the detection of Calbindin D-28K.
MAB1568
Anti-Calretinin Antibody, clone 6B8.2, 100 µg, Anti-Calretinin Antibody, clone 6B8.2 detects level of Calretinin & has been published & validated for use in IH & WB.
MAB377
Anti-NeuN Antibody, clone A60, 500 µg, Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB.
MAB1615
Anti-Neurofilament 70 kDa Antibody, clone DA2, 300 µL, This Anti-Neurofilament 70 kDa Antibody, clone DA2 is validated for use in WB, IC, IH for the detection of Neurofilament 70 kDa.
AB1989
Anti-Neurofilament H (200 kDa) Antibody, CT, 50 µL, Detect Neurofilament H (200 kDa) using this Anti-Neurofilament H (200 kDa) Antibody, C-terminus validated for use in IH & WB.
MAB318
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, 100 µL, Detect Tyrosine Hydroxylase using this Anti-Tyrosine Hydroxylase Antibody, clone LNC1 validated for use in IH, IH(P), IP & WB with more than 85 product citations.
10999644001 SP6/T7 Transcription Kit