EMAIL THIS PAGE TO A FRIEND
Oncogene
RUNX3 interacts with MYCN and facilitates protein degradation in neuroblastoma.
PMID 23851507
Abstract
RUNX3, a runt-related transcription factor, has a crucial role in dorsal root ganglion neurogenesis. Recent studies have suggested that RUNX3 acts as a tumor suppressor in stomach, colon and breast cancer. However, the biological role of RUNX3 in neuroblastoma remains elusive. Here we report that high levels of RUNX3 expression contribute to the favorable outcome in patients with neuroblastoma, whereas low levels of RUNX3 expression result in poor outcome. Array-based analysis suggested that the allelic loss at chromosome 1p36 is one of the reasons why expression of RUNX3 is downregulated in advanced neuroblastomas. Interestingly, the several patients survived from neuroblastoma with both high mRNA expressions of MYCN and RUNX3, suggesting that RUNX3 high expression might overcome the aggressive behavior of MYCN. Exogenous expression of RUNX3 strongly inhibits cell proliferation and migration in neuroblastoma cell lines. Furthermore, RUNX3 reduces the stability of MYCN protein in MYCN-amplified neuroblastoma cell lines, and this RUNX3-mediated MYCN degradation may depend on the physical interaction between RUNX3 and MYCN. Thus, our findings provide a tumor-suppressing mechanism by which RUNX3 inhibits the MYCN activity in neuroblastoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21184P Anti-c-Myc (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21222 Anti-c-Myc (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300319
Anti-MYC (Ab-358) antibody produced in rabbit, affinity isolated antibody
SAB4300320
Anti-MYC (Ab-373) antibody produced in rabbit, affinity isolated antibody
SAB4300318
Anti-MYC (Ab-58) antibody produced in rabbit, affinity isolated antibody
SAB2103448
Anti-MYC, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1409334
Anti-MYCN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410808
Anti-MYCN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101614
Anti-NOL3 antibody produced in rabbit, affinity isolated antibody
PRS2225
Anti-NOP30 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305017
ANTI-RUNX3 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405564
Anti-RUNX3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3300002
Monoclonal Anti-c-Myc antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous glycerol solution
WH0004609M2
Monoclonal Anti-MYC antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
WH0004613M1
Monoclonal Anti-MYCN antibody produced in mouse, clone 3H4, purified immunoglobulin, buffered aqueous solution
SAB1404101
Monoclonal Anti-MYCN, (N-terminal) antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
PLA0001
Rabbit anti-c-myc Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.