EMAIL THIS PAGE TO A FRIEND
Cancer letters
Antitumor activities of the targeted multi-tyrosine kinase inhibitor lenvatinib (E7080) against RET gene fusion-driven tumor models.
PMID 23856031
Abstract
RET gene fusions are recurrent oncogenes identified in thyroid and lung carcinomas. Lenvatinib is a multi-tyrosine kinase inhibitor currently under evaluation in several clinical trials. Here we evaluated lenvatinib in RET gene fusion-driven preclinical models. In cellular assays, lenvatinib inhibited auto-phosphorylation of KIF5B-RET, CCDC6-RET, and NcoA4-RET. Lenvatinib suppressed the growth of CCDC6-RET human thyroid and lung cancer cell lines, and as well, suppressed anchorage-independent growth and tumorigenicity of RET gene fusion-transformed NIH3T3 cells. These results demonstrate that lenvatinib can exert antitumor activity against RET gene fusion-driven tumor models by inhibiting oncogenic RET gene fusion signaling.