EMAIL THIS PAGE TO A FRIEND
Archives of virology
Telbivudine and adefovir combination therapy for patients with chronic lamivudine-resistant hepatitis B virus infections.
PMID 23857507
Abstract
We evaluated second-line salvage therapy with adefovir + telbivudine (group 1), adefovir followed by adefovir + telbivudine (group 2), or lamivudine + adefovir followed by adefovir + telbivudine (group 3) in hepatitis B patients with an inadequate virologic response to lamivudine treatment. Simple linear regression analysis showed that for each additional month of treatment, the most significant reduction in viral load occurred in group 1 (HBV DNA [Log10 IU/mL]: group 1, -0.149; group 2, -0.081; group 3, -0.123). Generalized estimating equation analysis revealed that compared to group 1, hepatitis B virus (HBV) DNA levels were 1.203 and 0.443 Log10 IU/mL higher in groups 2 and 3, respectively. Overall, a significant reduction in viral load (-0.060 Log10 IU/mL) was observed for each additional month of treatment. Adefovir + telbivudine treatment resulted in a significant reduction in HBV DNA levels. Moreover, telbivudine treatment resulted in a significant reduction in viral load (-0.050 Log10 IU/mL) compared to lamivudine treatment after the emergence of lamivudine resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0240
Adefovir, ≥98% (HPLC)
C8H12N5O4P
SML0293
Telbivudine, ≥98% (HPLC)
C10H14N2O5