EMAIL THIS PAGE TO A FRIEND
Journal of clinical pharmacology
A modified grapefruit juice eliminates two compound classes as major mediators of the grapefruit juice-fexofenadine interaction: an in vitro-in vivo "connect".
PMID 23878024
Abstract
The grapefruit juice (GFJ)-fexofenadine interaction involves inhibition of intestinal organic anion transporting polypeptide (OATP)-mediated uptake. Only naringin has been shown clinically to inhibit intestinal OATP; other constituents have not been evaluated. The effects of a modified GFJ devoid of furanocoumarins (~99%) and polymethoxyflavones (~90%) on fexofenadine disposition were compared to effects of the original juice. Extracts of both juices inhibited estrone 3-sulfate and fexofenadine uptake by similar extents in OATP-transfected cells (~50% and ~25%, respectively). Healthy volunteers (n = 18) were administered fexofenadine (120 mg) with water, GFJ, or modified GFJ (240 mL) by randomized, three-way crossover design. Compared to water, both juices decreased fexofenadine geometric mean AUC and C(max) by ~25% (P ≤ .008 and P ≤ .011, respectively), with no effect on terminal half-life (P = .11). Similar effects by both juices on fexofenadine pharmacokinetics indicate furanocoumarins and polymethoxyflavones are not major mediators of the GFJ-fexofenadine interaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9145
Estrone 3-sulfate potassium salt, ≥98%
C18H21KO5S