EMAIL THIS PAGE TO A FRIEND
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis
Relationship between neutrophil to lymphocyte ratio and slow coronary flow.
PMID 23892686
Abstract
We evaluated the relationship between neutrophil to lymphocyte ratio (NLR) and slow coronary flow (SCF). A total of 180 participants were recruited to the present study. Of all the participants, 82 patients were with SCF and 98 participants were with normal coronary arteries. Basal characteristics were recorded. Coronary flow was quantified by thrombolysis in myocardial infarction frame count. Basal characteristics were similar between the 2 groups. The NLR was significantly higher in the SCF group when compared to the control group (2.3 ± 0.8 and 1.5 ± 0.4 respectively, P < .001). In multiple logistic regression analysis, NLR remained as the independent predictor of SCF (P < 0.001). Our findings showed that NLR was significantly higher in the SCF group when compared to the control group with normal coronary arteries. We also showed that NLR was related to the presence of SCF rather than the extent of SCF. Besides these findings, we also showed the NLR as an independent predictor of SCF.