EMAIL THIS PAGE TO A FRIEND
Oral diseases
Cytokines induce MMP-3-regulated proliferation of embryonic stem cell-derived odontoblast-like cells.
PMID 23902456
Abstract
Matrix metalloproteinase (MMP)-3 expression increases after pulpectomy and accelerates angiogenesis in rat dental pulp by an uncharacterised mechanism. Odontoblasts, a major component of dental pulp, could represent a therapeutic target. We investigated whether MMP-3 activity is induced by cytokines and/or is associated with cell proliferation and apoptosis in embryonic stem cell-derived odontoblast-like cells. We used reverse transcriptase polymerase chain reaction, western blotting, an MMP-3 activity assay, a BrdU-cell proliferation enzyme-linked immunosorbent assay and DNA fragmentation analysis to evaluate siRNA-mediated downregulation of MMP-3 expression and activity, and any changes in the proliferative and apoptotic responses associated with this reduced expression. Pro-inflammatory cytokines (interleukin-1β, tumour necrosis factor-α and interferon-γ, at relatively low concentrations) induced MMP-3 mRNA and protein expression, and increased MMP-3 activity and cell proliferation, but not apoptosis. MMP-3 silencing produced a potent and significant suppression of cytokine-induced MMP-3 expression and activity, decreased cell proliferation and increased apoptosis. These effects were rescued by application of exogenous MMP-3. Our results suggest that pro-inflammatory cytokines induce MMP-3-regulated cell proliferation and anti-apoptosis effects in odontoblast-like cells derived from embryonic stem cells, in addition to their well-documented destructive role in inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501893
Anti-MMP3 (Cleaved-Phe100) antibody produced in rabbit, affinity isolated antibody
SAB1406132
Anti-MMP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007875
Anti-MMP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501702
Anti-TIMP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1409739
Anti-TIMP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4502971
Anti-TIMP1 antibody produced in rabbit, affinity isolated antibody
SAB1411349
Anti-TIMP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4502972
Anti-TIMP2 antibody produced in rabbit, affinity isolated antibody
SAB2106577
Anti-TIMP2 antibody produced in rabbit, affinity isolated antibody
EHU014871 MISSION® esiRNA, esiRNA targeting human MMP3 (esiRNA1)
EHU146741 MISSION® esiRNA, esiRNA human GAPDH (esiRNA1)
EMU031941 MISSION® esiRNA, esiRNA targeting mouse Mmp3 (esiRNA1)
EMU184491 MISSION® esiRNA, esiRNA targeting mouse Gapdh
SRP7783 MMP-3 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SAB5300422
Monoclonal Anti-MMP3 antibody produced in mouse, clone 4F10, purified immunoglobulin, buffered aqueous solution
SAB1402273
Monoclonal Anti-MMP3 antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
WH0007076M1
Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
SAB1404470 Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4E7, purified immunoglobulin, buffered aqueous solution
WH0007077M1
Monoclonal Anti-TIMP2 antibody produced in mouse, clone 5D7, purified immunoglobulin, buffered aqueous solution